Learn More
MSC SPI Plug Thread Gage, M5x0.8 Thread, 6H Class, Double End, Go & No Go
Supplier:  MSC 54334818
Encompass
This product is a Plug Thread Gage designed for inspecting internal threads. It features an M5x0.8 thread, 6H class, and is double-ended with both "Go" and "No Go" sides to ensure proper assembly of mating components.
  • Double-ended design with "Go" and "No Go" sides
  • M5x0.8 thread size for precise measurement
  • Constructed from durable tool steel for longevity
  • 6H class for accurate thread classification
  • Includes a handle for ease of use
  • Right hand thread direction
  • Inspection certificate included for traceability
  • Traceable to ASME B1.16M and NIST standards
  • Taperlock design facilitates inspection of internal threads
  • Ensures proper assembly of mating components
Catalog No. 50-005-28574