Learn More
MSC SPI Pocket Comparator, 10x, 5 Reticles, 1-3/8 in Lens Diameter
Supplier: MSC 03183852
This pocket comparator from SPI provides 10x magnification and features a 1-3/8 inch lens diameter. It comes equipped with 5 reticles, making it suitable for various inspection and measurement tasks. The unit includes a protective case for portability and storage.
- Includes a protective case
- Comes with 5 reticles/charts
- Offers 10x maximum magnification
- Features a 1-3/8 inch field of view