Learn More
MSC SPI Taperlock Thread Plug Gage, 3/8-16 Thread, 2B Class, Go Member
Supplier:  MSC 75888172
Encompass
This Taperlock Thread Plug Gage is designed for the inspection of internal threads, specifically featuring a 3/8-16 thread with a 2B class and functioning as a Go member. Constructed from durable tool steel, this single-end gage helps ensure the proper assembly of mating components and includes an inspection certificate traceable to NIST, conforming to ASME B1.2 standards.
  • Taperlock design.
  • Inspects internal threads for proper assembly.
  • Made from tool steel.
  • Single end configuration.
  • 3/8-16 thread size with 2B class.
  • Go pitch diameter of 0.3344 inch (8.4938 mm).
  • Right hand thread direction.
  • Conforms to ASME B1.2 and ANSI specification 1.16B.
  • Includes inspection certificate traceable to NIST.
  • Handle not included.
Catalog No. 50-005-30709