Learn More
MSC SPI Truncated Taperlock Thread Gage, #8-32 UNC, 2A, Go/No Go
Supplier:  MSC 46553293
Encompass
This truncated taperlock thread gage is designed for precise measurement of #8-32 UNC threads, featuring both "Go" and "No Go" functions for accurate gauging. Classified as 2A, it is ideal for setting adjustable thread ring gauges. Constructed from durable tool steel, it includes a handle for ease of use and conforms to ANSI/ASME B1.2 standards, ensuring reliability and accuracy.
  • Designed for #8-32 UNC threads
  • Features both "Go" and "No Go" functions
  • Classified for 2A thread fit
  • Made from durable tool steel
  • Includes a convenient handle
  • Conforms to ANSI/ASME B1.2 standards
  • Includes an inspection certificate traceable to NIST
  • Offers X tolerance on pitch diameter
  • Provides W tolerance on flank angles and lead
Catalog No. 50-005-27799