Learn More
MSC SPI V-Block, Magnetic Hardened Steel, 1-3/4 in Capacity, 90° V Angle
Supplier: MSC 07176845
The SPI V-Block is designed for precise measurement and inspection tasks in laboratory and industrial settings. Made from hardened steel, it offers durability and accuracy, making it suitable for holding cylindrical workpieces.
- Magnetic functionality for secure workpiece holding.
- Hardened steel construction ensures durability and stability.
- Maximum capacity of 1-3/4 inches for versatile applications.
- 90-degree V angle supports various cylindrical component sizes.
- High precision with V parallelism of 5.00 micrometers.
- Hardness of 55 HRC for long-lasting performance.
- Suitable for inspection applications.