Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike Trimer (T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681R, D950N), His,Avitag™
Supplier:  ACROBiosystems SPNC82EI25UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ei) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 139.5 kDa.
Catalog No. NC2010477