Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 22 x 8, Length x Dia. in inches: 7/8 x 5/16
SDP
Supplier:  Chemglass Life Sciences CG200106
Encompass_Preferred
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 22 x 8, Length x Dia. in inches: 5/8 x 5/16. Octagon cross-section having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.SEE CG-2001-50 FOR STIR BAR KIT
Catalog No. CG200106