Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 76.2 x 12.7, Length x Dia. in inches: 3 x 1/2
SDP
Supplier:  Chemglass Life Sciences CG200120
Encompass_Preferred
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 76.2 x 12.7, Length x Dia. in inches: 3 x 1/2. Octagon cross-section having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.SEE CG-2001-50 FOR STIR BAR KIT
Catalog No. CG200120