Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Glass, Length x Dia. in mm: 25.4 x 9.52, Length x Dia. in inches: 1 x 3/8
SDP
Supplier:  Chemglass Life Sciences CG200412
Encompass_Preferred
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 25.4 x 9.52, Length x Dia. in inches: 1 x 3/8. Borosilicate covered Alnico magnet for use in high temperature reactions.
Catalog No. 50-872-472