Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, "V" Shaped, 9mm Height x 5.5mm Diameter
SDP
Supplier:  Chemglass Life Sciences CG200810
Encompass_Preferred
Stir Bar, Magnetic, PTFE, "V" Shaped, 9mm Height x 5.5mm Diameter. "V" Shaped PTFE magnetic stir bar. Design permits rotation in small test tubes and conical vessels such as MW-80 reaction vials. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. CG200810