Learn More
Chemglass Life Sciences Spinner Flask Only, 6000mL, Jacketed, BioProcess, Plain Bottom
SDP
Supplier:  Chemglass Life Sciences CLS1430J6L1
Encompass_Preferred
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask, manufactured from Type 1 Class A borosilicate glass.Flasks are typically used with a circulating water bath.Jacket volumes are approximate and flasks are not graduated.Note: Paddle and Cap assembly are not included.
Catalog No. 50-273-3290