Learn More
Medchemexpress LLC Spiraeoside | 20229-56-5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN825310MG
Encompass_Preferred
Spiraeoside is an orally active natural compound that exhibits antioxidant activity by inhibiting reactive oxygen species (ROS) and malondialdehyde production. It also possesses antiallergic, anti-inflammatory, and antitumor activities.
  • Inhibits reactive oxygen species (ROS) and malondialdehyde production.
  • Acts as an antiallergic, anti-inflammatory, and antitumor agent.
  • Elevates p-Akt, nuclear Nrf2, and HO-1 expression in AC16 cells.
  • Protects AC16 cells from high glucose-induced oxidative stress, cell injury, and apoptosis.
  • Activates the PI3K/Akt/Nrf2 pathway.
  • Demonstrates ulcer preventive ability in male Wistar rats.
Catalog No. 50-004-19444