Learn More
Medchemexpress LLC Spiraeoside | 20229-56-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN82535MG
Encompass_Preferred
Spiraeoside is an orally active natural compound that exhibits antioxidant activity by inhibiting reactive oxygen species (ROS) and malondialdehyde production. It also possesses antiallergic, anti-inflammatory, and antitumor activities. It protects human cardiomyocytes against high glucose-induced injury, oxidative stress, and apoptosis by activating the PI3K/Akt/Nrf2 pathway.
  • Exhibits antioxidant activity
  • Inhibits reactive oxygen species (ROS) and malondialdehyde production
  • Possesses antiallergic, anti-inflammatory, and antitumor activities
  • Protects human cardiomyocytes against high glucose-induced injury, oxidative stress, and apoptosis
  • Activates the PI3K/Akt/Nrf2 pathway
Catalog No. 50-004-19811