Learn More
Medchemexpress LLC HY-100971 5mg Medchemexpress, Spiramide CAS:510-74-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009715MG
Encompass_Preferred
Medchemexpress, HY-100971 5mg Spiramide CAS:510-74-7 Spiramide (AMI-193) is a potent and selective antagonist of 5-HT2 and dopamine D2 receptor, with Kis of 2 nM and 3 nM, respectively. Spiramide has >2000-fold selectivity for 5-HT2 versus 5-HT1C (Ki=4300 nM) receptors. Spiramide exhibits antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0611