Learn More
Medchemexpress LLC Spiro[2H-1-benzothiopyran-3(4H),1'-cyclopropan]-4-one, 2'-(4-methoxyphenyl)-, (1'R,2'R)-rel- | 1799392-31-6 | 99.8% | 296.38 g/mol | C18H16O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1415915MG
Encompass_Preferred
CYT296 is a small-molecule chromatin de-condensation compound for research use. It promotes an open chromatin state and can improve induction of induced pluripotent stem cells (iPSCs) when used with defined transcription factors.
  • Promotes chromatin de-condensation and open chromatin state.
  • Enhances efficiency of iPSC induction when combined with defined factors.
  • Provided as a high-purity research chemical (99.8%).
  • Available in small vial pack sizes suitable for laboratory experiments.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-22792