Learn More
Medchemexpress LLC Spiro[3H-indole-3,4'-[4H]thiopyran]-5-carboxamide, N-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]methyl]... | 1631164-24-3 | 99.5% | 672.11 g/mol | C29H26ClF4N3O5S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1330805MG
Encompass_Preferred
BAY-784 is a small-molecule gonadotropin releasing hormone receptor (GnRH-R) antagonist used as a research probe to inhibit GnRH-mediated signaling in vitro and in vivo. It is characterized by nanomolar potency, defined physicochemical properties, and high chemical purity for reliable assay performance.
  • Small-molecule GnRH receptor antagonist with nanomolar potency.
  • Reported IC50: 21 nM (human), 24 nM (rat).
  • Molecular formula C29H26ClF4N3O5S2; molecular weight 672.11 g/mol.
  • High chemical purity (99.5%).
  • Supplied as a 5 mg research pack for assay and pharmacology studies.
  • Useful for pharmacology, receptor biology, and signaling research.
Catalog No. 50-002-62274