Learn More
Medchemexpress LLC Spironolactone | 52-01-7 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB05615G
Encompass_Preferred
Spironolactone is a high-purity chemical compound, with 99.8% purity, primarily acting as an aldosterone antagonist and also demonstrating antagonist activity at the androgen receptor. This compound is offered in solid form, soluble at 10 mM in DMSO, and is notable for its involvement in podocyte autophagy and pain regulation, alongside its multifaceted antihypertensive effects.
  • Improves hypertension-related vascular hypertrophy and remodeling.
  • Reduces aldosterone-induced vascular and soft tissue calcification.
  • Alleviates vascular dysfunction in type II diabetic mice.
  • Promotes podocyte autophagy and regulates pain.
  • Acts as an androgen receptor antagonist for hormonal regulation.
Catalog No. 50-004-07848