Learn More
Medchemexpress LLC Spironolactone-d7 | 99.96% | 100 UG
SDP
Supplier:  Medchemexpress LLC HYB0561S100UG
Encompass_Preferred
Spironolactone-d7 is the deuterium labeled version of Spironolactone, an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM and a potent antagonist of androgen receptor with an IC50 of 77 nM. It is known to promote autophagy in podocytes and is commonly used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound for accurate tracing
  • Can be used as an internal standard in quantitative analysis
  • High purity (99.96%) for reliable results
  • Supports research in drug development and metabolism
  • Applicable in studies related to cancer and endocrine diseases
Catalog No. 50-004-08648