Learn More
Medchemexpress LLC Spironolactone (Standard) | 52-01-7 | 98.55% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0561R
Encompass_Preferred
Spironolactone is an orally active analytical standard intended for research and analytical applications. It functions as an aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM, and also acts as a potent androgen receptor antagonist with an IC50 of 77 nM. Additionally, it has been shown to promote autophagy in podocytes.
  • Orally active.
  • Aldosterone mineralocorticoid receptor antagonist.
  • Androgen receptor antagonist.
  • Promotes autophagy in podocytes.
  • Used in qualitative, quantitative and methodological research experiments.
Catalog No. 50-004-50742