Learn More
Medchemexpress LLC Spironolactone (Standard) | 52-01-7 | 98.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0561R100MG
Encompass_Preferred
Spironolactone (Standard) serves as the analytical standard for Spironolactone. It is designed for research and analytical applications. Spironolactone (SC9420) is an orally active antagonist of the aldosterone mineralocorticoid receptor, with an IC50 of 24 nM. It also acts as a potent antagonist of the androgen receptor, exhibiting an IC50 of 77 nM. Furthermore, Spironolactone has been shown to promote autophagy in podocytes.
  • Intended for research and analytical applications
  • Analytical standard for Spironolactone
  • Orally active aldosterone mineralocorticoid receptor antagonist (IC50 of 24 nM)
  • Potent antagonist of androgen receptor (IC50 of 77 nM)
  • Promotes autophagy in podocytes
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-08430