Learn More
Medchemexpress LLC Spiroxamine | 118134-30-8 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HY12188410MM 1ML
Spiroxamine is an analytical-grade reference standard of the fungicide spiroxamine (CAS 118134-30-8) intended for research and analytical use. It is supplied at high purity and is available as a 10 mM solution in DMSO (1 mL) or as solid milligram quantities for analytical workflows.
- High purity suitable for analytical applications.
- Available as 10 mM solution in DMSO (1 mL) and as solid mg quantities.
- Molecular formula C18H35NO2; molecular weight 297.48 g·mol⁻1.
- Intended for research and analytical testing; not for human therapeutic use.
- Supplied to support reconstitution, assay preparation, and reference analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.