Learn More
Medchemexpress LLC HY-111360 5mg Medchemexpress, SPL-707 CAS:2195361-33-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1113605MG
Encompass_Preferred
Medchemexpress, HY-111360 5mg SPL-707 CAS:2195361-33-0 SPL-707 is an orally active, selective signal peptide peptidase-like 2a (SPPL2a) inhibitor with an IC50 of 77 nM for hSPPL2a. SPL-707 inhibits γ-secretase (IC50=6.1 μM) and SPP (IC50=3.7 μM). SPL-707 has the potential for autoimmune diseases research by targeting B cells and dendritic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0789