Learn More
Meso Scale Discovery SPLEX HUM IL12p70 Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151G3S4
Encompass
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay for measuring human interleukin 12 (IL-12) in various samples It uses electrochemiluminescence (ECL) detection offering high sensitivity a wide dynamic range and low sample use The assay is specific for IL-12p70 with minimal cross-reactivity The kit includes a 96-well plate antibodies buffers diluent and calibrators from 3 2 to 10 000 pg/mL The assay takes 3 5 hours including a 2-hour incubation
Catalog No. 50-000-80186
Only null left
Add to Cart
Request a Quote