Learn More
Meso Scale Discovery SPLEX HUM Neurofilament L Kit
SDP
Supplier:  Meso Scale Discovery K151AKGS1
Encompass
The S-PLEX Human Neurofilament L Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of neurofilament light chain (NfL) in human serum plasma and CSF The assay uses MSD s proprietary S-PLEX technology is highly sensitive has a broad dynamic range of 47 - 10 000 pg/mL and shows no cross-reactivity with neurofilament medium and heavy chains The kit includes a 96-well plate coated with anti-NfL capture antibody detection antibody read buffer sample diluent calibrators and controls The assay takes 3 5 hours total time and requires 10 L sample volume
Catalog No. 50-000-78216