Learn More
Meso Scale Discovery SPLEX Proinflam PNL human
SDP
Supplier:  Meso Scale Discovery K15396S4
Encompass
The Meso Scale Discovery S-Plex Proinflammatory Panel 1 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 proinflammatory biomarkers in human serum plasma and cell culture supernatants The kit includes a 10-spot MULTI-ARRAY 96-well plate pre-coated with capture antibodies for the following analytes IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- The assay uses electrochemiluminescence for high sensitivity and a broad dynamic range It requires 50 L or less sample volume per well The kit includes all necessary reagents and can perform up to 94 assays in duplicate It is for research use only
Catalog No. 50-000-82250
Only null left
Add to Cart
Request a Quote