Learn More
Meso Scale Discovery SPLEX SARSCoV2 N Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K150ADHS4
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 in human serum plasma and saliva This assay utilizes electrochemiluminescent detection technology on the SECTOR Imager 2400 platform The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium assay diluent wash buffer sample diluent and calibrators This is an in vitro diagnostic test for the presumptive qualitative detection of SARS-CoV-2 antibodies
Catalog No. 50-000-80526
Only null left
Add to Cart
Request a Quote