Learn More
Meso Scale Discovery SPLEX SARSCoV2 Spike KitQP
SDP
Supplier:  Meso Scale Discovery K150ADJS24
Encompass
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that detects antibodies to the SARS-CoV-2 spike protein in human serum and plasma This qualitative assay uses MSD s proprietary SECTOR technology is compatible with the MESO QuickPlex SQ 120 instrument and provides results in under 2 hours The kit includes a 96-well plate coated with recombinant SARS-CoV-2 spike protein assay diluent calibrators wash buffer read buffer and plate seals This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81762
Only null left
Add to Cart
Request a Quote