Learn More
Thermo Scientific Vanquish Split Sampler Sample Loops | N/A | 1 EA
Supplier:  Thermo Scientific 6252.1940
Encompass
Automate sample injection into your Thermo Scientific Vanquish HPLC or UHPLC system with Thermo Scientific Vanquish Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 L sizes to meet your application requirements. Thermo Scientific Viper Fittings are included with our sample loops to make them easy to install with leak-free connections.
Catalog No. NC2170274