Learn More
Medchemexpress LLC SPOP-IN-6b dihydrochloride | 99.7% | 573.51 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY122615B10MG
Encompass_Preferred
SPOP-IN-6b dihydrochloride is a potent speckle-type POZ protein (SPOP) inhibitor with an IC50 of 3.58 μM. It is intended for research use only and not for sale to patients.
  • Potent SPOP inhibitor with an IC50 of 3.58 μM.
  • Suppresses SPOP activity, leading to an increase in suppression cancer substrate proteins PTEN and DUSP7 content in vitro (IC50s of 2-10.2 μM for A498, Caki-2, Ketr-3, 769-P, 0S-RC-2, and 786-0 cells).
  • Slows down tumor growth in nude mice by suppressing SPOP (40-80 mg/kg; intraperitoneal injection; daily; for 25 days).
Catalog No. 50-004-37201