Learn More
TRUE INDICATING SPORE THREAD GEO STEARO 100/PK
Supplier:  TRUE INDICATING TT06
Encompass_Preferred
Geobacillus stearothermophilus Cell line 7953 Spore Thread for Monitoring Steam, 32 mm X 0.1 mm diameter thread, Population level of 106/ threadindividually packaged in glassine envelopes 30 mm x 38 mm100 Threads/PackISO 11138-1 and 11138-3 compliant
Catalog No. NC2295465