Learn More
Proteintech Group Inc Spot-Cap™ and Peptide
SDP
Supplier:  Proteintech Group Inc ECAEP
Encompass
Spot-Cap is a Nanobody-based affinity resin optimized for one-step protein purification of Spot-tagged proteins.
Catalog No. 50-225-8172