Learn More
Maine Molecular Quality Controls Inc SPOTFIRE RSP Negative Control
SDP
Catalog No. NC3886287
Encompass
SPOTFIRE RSP Negative Control is intended for use (as applicable) as an external negative assayed quality control to monitor performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Bordetella parapertussis, Bordetella pertussis, Chlamydia pneumoniae, Mycoplasma pneumoniae, Streptococcus dysgalactiae (Group C/G Strep), Streptococcus pyogenes (Group A Strep), Adenovirus, Coronavirus SARS-CoV-2, Coronavirus (seasonal), Human Metapneumovirus, Human Rhinovirus/Enterovirus, Influenza A Subtype H1-2009, Influenza A Subtype H3, Influenza B, Parainfluenza Virus and Respiratory Syncytial Virus using the BIOFIRE SPOTFIRE Respiratory (R) Panel, BIOFIRE SPOTFIRE Respiratory (R) Panel Mini, BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel, and BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel Mini on the BIOFIRE SPOTFIRE System.
Catalog No. NC3886287 Supplier Maine Molecular Quality Controls Inc Supplier No. M42738
SPOTFIRE RSP Negative Control is intended for use (as applicable) as an external negative assayed quality control to monitor performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Bordetella parapertussis, Bordetella pertussis, Chlamydia pneumoniae, Mycoplasma pneumoniae, Streptococcus dysgalactiae (Group C/G Strep), Streptococcus pyogenes (Group A Strep), Adenovirus, Coronavirus SARS-CoV-2, Coronavirus (seasonal), Human Metapneumovirus, Human Rhinovirus/Enterovirus, Influenza A Subtype H1-2009, Influenza A Subtype H3, Influenza B, Parainfluenza Virus and Respiratory Syncytial Virus using the BIOFIRE SPOTFIRE Respiratory (R) Panel, BIOFIRE SPOTFIRE Respiratory (R) Panel Mini, BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel, and BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel Mini on the BIOFIRE SPOTFIRE System.