Learn More
Grainger Spotting Drill Bit Carbide 1
Supplier:  Grainger 16R621
Encompass_Preferred
The Micro 100 1" Spotting Drill Bit is a high-speed steel spotting drill bit with a 1" cutting diameter and 1/2" shank diameter. It has a 118 degree split point tip angle and is designed for use on steel, cast iron, aluminum, and other non-ferrous metals. The bit features a TiN coating for increased wear resistance and lubricity. It can be used for centering, spotting, and drilling applications at speeds up to 2500 RPM with proper coolant. Made in USA by Micro 100, this is a high quality spotting drill bit well-suited for precision machining applications.
Catalog No. 50-276-5289