Learn More
Aviva Systems Biology Corp SPPL2B Antibody - N-terminal region (ARP44989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPPL2B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SPPL2B. Alternate names: IMP4, KIAA1532, MGC111084, PSL1, IMP-4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58949