Learn More
Apexbio Technology LLC SPQ 83907-40-8 100mg
SDP
Supplier:  Apexbio Technology LLC A4671100
Encompass_Preferred
SPQ (CAS 83907-40-8) is a fluorescent probe used to monitor membrane-associated ion transport events It is designed to facilitate the detection of dynamic activities of membrane-bound ion channels and transport proteins by measuring changes in fluorescence intensity governed by ion-sensitive quenching mechanisms SPQ exerts its functional utility through ionic concentration-dependent fluorescence quenching enabling researchers to directly assess ion flux across cellular or subcellular membranes Based on these properties SPQ holds research potential in studies of membrane physiology transporter protein functionality and ion regulatory pathways in cell biology and pharmacological research
Catalog No. 50-000-94103