Learn More
Medchemexpress LLC SPQ | 83907-40-8 | 98.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD093610MM/1ML
Encompass_Preferred
SPQ is a fluorescent dye designed for measuring chloride ion concentration transients and transport rates in biological systems such as vesicles, cells, and intact epithelium. It accurately reports real-time changes in intracellular chloride concentration, making it suitable for various research applications.
  • Measures chloride ion concentration transients
  • Measures transport rates in vesicles, cells, and intact epithelium
  • Accurately reports real-time changes in intracellular chloride concentration
  • Suitable for cell staining analysis
  • Functions as an ion fluorescent probe
Catalog No. 50-004-10084