Learn More
Medchemexpress LLC SPQ | 83907-40-8 | 98.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0936100MG
Encompass_Preferred
SPQ is a fluorescent dye used to measure chloride ion concentration transients and transport rates in vesicles, cells, and intact epithelium.
  • Measures chloride ion concentration transients.
  • Measures transport rates in vesicles, cells, and intact epithelium.
  • Accurately reports changes in intracellular chloride concentration.
Catalog No. 50-004-09608