Learn More
ABclonal Technology SPR Rabbit pAb
SDP
Supplier:  ABclonal Technology A11694100UL
Encompass_Preferred
This gene encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Catalog No. 50-001-16410