Learn More
Essendant Spray Bottle, 32 oz, Clear, 24/Case
Supplier:  Essendant BWK00032
Encompass_Preferred
Bottle only. Spray trigger sold separately. Convenient bottles uses an embossed scale to encourage users to properly proportion chemicals. Tapered neck provides a more comfortable grip when using with a trigger sprayer. Easy-to-use, multi-purpose bottle is a great choice for everyday cleaning tasks. Material(s): HDPE; Color(s): Clear; Capacity (Volume): 32 oz.
Catalog No. NC1733082