Learn More
US Plastic Corporation 28/400 Green & White Sprayer with 7-1/4" Dip Tube
SDP
Supplier:  US Plastic Corporation 66854
Encompass
28/400 Green & White Sprayer with 7-1/4" Dip Tube (Bottle Sold Separately). As the leading general-purpose trigger sprayer in the janitorial and industrial market, this sprayer produces up to 40% more output per stroke, significantly reducing the number of times you pull the trigger. The rubber piston cup extends the life and provides broader chemical resistance, while the outer body protects the piston assembly to prevent damage during use and storage. The high output and extra-large spray pattern make quick work of any job.
Catalog No. NC1865156