Learn More
Agilent Technologies SPRAY CHAMBER DRAIN TUBE ASSY
Supplier:  Agilent Technologies G328060555
Encompass
Spray chamber drain tube assembly for Quartz Scott double-pass. Suits Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS. Drain tube with push-fit connection to spray chamber drain and barbed connector to peristaltic pump drain tubing
Catalog No. NC2188013