Learn More
Essendant Rust-Oleum Hard Hat High Performance V2100 System Rust Preventive Enamel Spray Paint, Gloss Black, 15 oz Aerosol Can, 6/Carton
Supplier:  Essendant RSTV2179838CT
Encompass_Preferred
Hard Hat High Performance V2100 System Rust Preventive Enamel Spray Paint provides superior rust protection and a top-quality finish for a variety of industrial projects As a high performance oil-based enamel paint it s particularly well suited for structural steel handrails industrial equipment machinery pipes storage tanks and more Advanced Hard Hat V2100 System delivers the best-possible corrosion protection in an easy one-coat coverage format while simultaneously ensuring excellent chip crack and peel resistance Extra-wide comfort nozzle reduces finger fatigue and enables spraying from any angle while a high-output spray tip and fast dry times help users complete projects more quickly Enamel withstands temperatures up to 200 F Two-year rust-proof guarantee Paint Sub-Category- Enamel Oil-Based Spray Paint Intended Application Environment- Interior/Exterior Finish/Sheen- Gloss Color(s)- Black
Catalog No. 50-000-26954