Learn More
ABclonal Technology SPTLC2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A117161000UL
Encompass_Preferred
This gene encodes a long chain base subunit of serine palmitoyltransferase. Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine. Mutations in this gene were identified in patients with hereditary sensory neuropathy type I.
Catalog No. 50-001-14074