Learn More
Grainger Spud Coupling Assbly American Standard
Supplier:  Grainger 16U024
Encompass_Preferred
The American Standard Spud Coupling Assembly is a 1-1/2" spud coupling assembly that fits 1-1/2" copper tubing. It is made of chrome-plated brass and has a maximum pressure rating of 125 psi at 70°F. The assembly includes a coupling nut, gasket, and tailpiece. It can be used to connect copper tubing to fixtures and valves in plumbing applications. The coupling has a female NPT thread on one end to connect to the fixture or valve, and a nut and gasket on the other end to connect to the copper tubing. This spud coupling provides a secure connection from copper tubing to threaded plumbing fittings.
Catalog No. 50-276-7629