Learn More
AdipoGen SPulegone
SDP
Supplier:  AdipoGen CDXP0508M050
Encompass_Preferred
Chemical. CAS 3391-90-0. Formula C10H16O. MW BD9837. S-Pulegone is a monoterpene utilized as a particularly actractive chiral starting material for the synthesis of more complex natural products. This compound has also shown to have antimalarial properties.
Catalog No. 50-256-0180