Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Veco Square Center Reference 200 mesh Gold Grid
SDP

Supplier:  Electron Microscopy Sciences 0200AU

Encompass_Preferred

Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm

Catalog No. 50-980-113


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.