Learn More
Grainger Sq. End Mill Single End Carb 0.0130
Supplier:  Grainger 14M757
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. The square end measures 0.5 inches across. This end mill is designed for use on steel, cast iron, aluminum, and other non-ferrous metals. It can be run at speeds up to 2000 SFM when taking light cuts.
Catalog No. 50-275-2040