Learn More
Grainger Sq. End Mill Single End Carb 0.60mm
Supplier:  Grainger 14K545
Encompass_Preferred
The Grainger 14K545 is a Westward brand 1/4-inch drive click-type torque wrench with a range of 10 to 150 inch pounds. It features a reversible ratchet head, knurled grip, and a positive lock with torque adjustment knob. This click-type wrench emits an audible click that can be heard and felt when the preset torque value is reached, allowing for accurate tightening. The tool has a length of 15.5 inches and weighs 1.15 pounds. It is made in the USA and comes with a certificate of calibration traceable to NIST standards. The wrench is ideal for precise tightening applications.
Catalog No. 50-275-3307