Learn More
Grainger Sq. End Mill Single End Carb 0.0240
Supplier:  Grainger 14K506
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and is designed for general purpose milling in most material. The end mill's composition is M2 high speed steel which provides good wear resistance and allows higher cutting speeds than regular carbon steel end mills.
Catalog No. 50-275-1713