Learn More
Grainger Sq. End Mill Single End Carb 0.0100
Supplier:  Grainger 14K477
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank reduction. It has an American National standard taper shank and is designed for general purpose milling in most material. The end mill's peripheral cutting edges can mill axially and interpolate.
Catalog No. 50-275-3197